Assay Design
Gene Transcripts
- Gene Symbol
- Tep1
- Entrez Gene ID
-
34937
- Gene Name
- Thioester-containing protein 1
- Gene Aliases
-
BG:DS00365.2, CG18096, Dmel\CG18096, Dmel_CG18096, TEP I, TEP1, TEP1mel, Tep 1, Tep I, Tep-1, TepI, dTEPI, tep1
- Chromosome Location
-
Chr.2L: 15888640 - 15893811 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041183
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078854.2 |
NP_523578.1 |
6 - 7 |
1910 |
72 |
GenBank mRNA |
AJ269538.1 |
- |
6 - 7 |
1910 |
72 |
View Full Product Details