Assay Design



Gene Transcripts

Gene Symbol
yellow-b
Entrez Gene ID
35005
Gene Name
CG17914-PA
Gene Aliases
CG17914, Dmel\CG17914, Dmel_CG17914, Yellow-b, yell-b
Chromosome Location
Chr.2L: 16754336 - 16758283 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032601
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299058.1 NP_001285987.1 2 - 3 528 95
NM_078862.2 NP_523586.1 2 - 3 348 95
GenBank mRNA AF237613.1 - 2 - 3 312 95
AY118575.1 - 2 - 3 312 95

View Full Product Details