Assay Design



Gene Transcripts

Gene Symbol
Dhc36C
Entrez Gene ID
35061
Gene Name
Dynein heavy chain at 36C
Gene Aliases
CG5526, DHC, DHC7A, Dhc, Dhc36c, Dmel\CG5526, Dmel_CG5526, dhc
Chromosome Location
Chr.2L: 17501401 - 17525515 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013810
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382179.1 NP_001369131.1 1 - 2 603 73
NM_078867.4 NP_523591.2 1 - 2 603 73

View Full Product Details