Assay Design
Gene Transcripts
- Gene Symbol
- Or42b
- Entrez Gene ID
-
35516
- Gene Name
- Odorant receptor 42b
- Gene Aliases
-
41E.1, 42b, CG12754, DOR118, Dmel\CG12754, Dmel_CG12754, OR42b
- Chromosome Location
-
Chr.2R: 5797195 - 5798512 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033043
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078900.2 |
NP_523624.2 |
1 - 2 |
768 |
78 |
View Full Product Details