Assay Design
Gene Transcripts
- Gene Symbol
- Cyp9b1
- Entrez Gene ID
-
35634
- Gene Name
- Cytochrome P450-9b1
- Gene Aliases
-
9b1, CG4485, CYP9B1, Dmel\CG4485, Dmel_CG4485, P-450, P450
- Chromosome Location
-
Chr.2R: 7127838 - 7129808 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015038
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078921.4 |
NP_523645.1 |
1 - 2 |
237 |
154 |
View Full Product Details