Assay Design
Gene Transcripts
- Gene Symbol
- Or47b
- Entrez Gene ID
-
36212
- Gene Name
- Odorant receptor 47b
- Gene Aliases
-
47E.2, 47b, CG13206, DOR25, DOR47E.2, DmOr47b, Dmel\CG13206, Dmel_CG13206, OR 47b, OR47b, Or47E.2, or47b
- Chromosome Location
-
Chr.2R: 11319710 - 11321312 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026385
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078966.3 |
NP_523690.3 |
3 - 4 |
825 |
60 |
View Full Product Details