Assay Design



Gene Transcripts

Gene Symbol
Buffy
Entrez Gene ID
36251
Gene Name
48A-E Bcl-2-like
Gene Aliases
BUFFY, Bcl-2-48AE, Buffy/Borg2, Buffy/DBorg-2, Buffy/dBorg-2, CG8238, Dbx, Dmel\CG8238, Dmel_CG8238, Drob2, buffy, dBORG-2/BUFFY, dBorg-2, dBorg2, dbuffy
Chromosome Location
Chr.2R: 11679492 - 11688557 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040491
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078978.2 NP_523702.1 2 - 3 413 80
GenBank mRNA AY075219.1 - 2 - 3 432 80

View Full Product Details