Assay Design
Gene Transcripts
- Gene Symbol
- Buffy
- Entrez Gene ID
-
36251
- Gene Name
- 48A-E Bcl-2-like
- Gene Aliases
-
BUFFY, Bcl-2-48AE, Buffy/Borg2, Buffy/DBorg-2, Buffy/dBorg-2, CG8238, Dbx, Dmel\CG8238, Dmel_CG8238, Drob2, buffy, dBORG-2/BUFFY, dBorg-2, dBorg2, dbuffy
- Chromosome Location
-
Chr.2R: 11679492 - 11688557 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040491
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078978.2 |
NP_523702.1 |
3 - 4 |
563 |
85 |
GenBank mRNA |
AY075219.1 |
- |
3 - 4 |
582 |
85 |
View Full Product Details