Assay Design
Gene Transcripts
- Gene Symbol
- Or49b
- Entrez Gene ID
-
36413
- Gene Name
- Odorant receptor 49b
- Gene Aliases
-
49D.1, 49b, AN13, CG17584, DOR105, Dmel\CG17584, Dmel_CG17584, Or-49b
- Chromosome Location
-
Chr.2R: 12842665 - 12844704 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028963
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078997.3 |
NP_523721.1 |
1 - 2 |
906 |
84 |
GenBank mRNA |
BT024438.1 |
- |
1 - 2 |
906 |
84 |
View Full Product Details