Assay Design



Gene Transcripts

Gene Symbol
Roc2
Entrez Gene ID
36246
Gene Name
Regulator of cullins 2
Gene Aliases
BcDNA:RE61847, CG8998, Dmel\CG8998, Dmel_CG8998, Roc, TRET, dRoc2, roc2
Chromosome Location
Chr.2R: 11652756 - 11679354 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0044020
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169648.2 NP_001163119.1 3 - 4 434 71
NM_136847.3 NP_610691.1 2 - 3 338 71
GenBank mRNA AY113544.1 - 2 - 3 336 71
BT072910.1 - 3 - 4 393 71

View Full Product Details