Assay Design
Gene Transcripts
- Gene Symbol
- ClC-b
- Entrez Gene ID
-
36381
- Gene Name
- Chloride channel-b
- Gene Aliases
-
CG8594, Clcn7, DmClC-b, DmClCb, Dmel\CG8594, Dmel_CG8594
- Chromosome Location
-
Chr.2R: 12572068 - 12575299 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033755
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136954.4 |
NP_610798.1 |
5 - 6 |
1762 |
85 |
GenBank mRNA |
AY071581.1 |
- |
5 - 6 |
1718 |
85 |
View Full Product Details