Assay Design
Gene Transcripts
- Gene Symbol
- ppk3
- Entrez Gene ID
-
246504
- Gene Name
- pickpocket 3
- Gene Aliases
-
CG15872, CG30181, Dmel\CG30181, Dmel_CG30181, cg30181
- Chromosome Location
-
Chr.2R: 23538631 - 23540773 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050181
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166606.1 |
NP_726336.1 |
1 - 2 |
688 |
76 |
View Full Product Details