Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169803.2 | NP_001163274.1 | 15 - 16 | 2361 | 75 |
NM_001169804.2 | NP_001163275.2 | 11 - 12 | 2521 | 75 | |
NM_001169805.2 | NP_001163276.1 | 14 - 15 | 2188 | 75 | |
NM_001169807.2 | NP_001163278.2 | 14 - 15 | 2474 | 75 | |
NM_137970.5 | NP_611814.3 | 15 - 16 | 2192 | 75 | |
NM_166616.3 | NP_726350.1 | 14 - 15 | 2474 | 75 | |
NM_166617.2 | NP_726351.1 | 12 - 13 | 1608 | 75 | |
NM_166618.3 | NP_726352.1 | 13 - 14 | 1957 | 75 | |
NM_166619.2 | NP_726353.1 | 2 - 3 | 481 | 75 | |
GenBank mRNA | AY060327.1 | - | 2 - 3 | 481 | 75 |
BT011336.1 | - | 14 - 15 | 2438 | 75 | |
BT021326.1 | - | 15 - 16 | 2188 | 75 | |
BT133111.1 | - | 13 - 14 | 1957 | 75 |