Assay Design
Gene Transcripts
- Gene Symbol
- yki
- Entrez Gene ID
-
37851
- Gene Name
- yorkie
- Gene Aliases
-
CG4005, Dmel\CG4005, Dmel_CG4005, YAP, YKI, Yki, yap
- Chromosome Location
-
Chr.2R: 24065975 - 24068485 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034970
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138035.3 |
NP_611879.4 |
2 - 3 |
1111 |
88 |
View Full Product Details