Assay Design



Gene Transcripts

Gene Symbol
yki
Entrez Gene ID
37851
Gene Name
yorkie
Gene Aliases
CG4005, Dmel\CG4005, Dmel_CG4005, YAP, YKI, Yki, yap
Chromosome Location
Chr.2R: 24065975 - 24068485 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034970
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043103.2 NP_001036568.2 4 - 5 1319 72
NM_001363931.1 NP_001350857.1 4 - 5 1319 72
NM_138035.3 NP_611879.4 3 - 4 1163 72
NM_166657.5 NP_726414.3 5 - 6 1254 72
GenBank mRNA BT132826.1 - 4 - 5 1240 72
DQ099897.1 - 4 - 5 1319 72

View Full Product Details