Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001259573.1 | NP_001246502.1 | 6 - 7 | 1968 | 56 |
NM_138121.2 | NP_611965.2 | 7 - 8 | 2031 | 56 | |
NM_166686.1 | NP_726481.1 | 6 - 7 | 1968 | 56 | |
NM_166687.1 | NP_726482.1 | 6 - 7 | 1968 | 56 | |
GenBank mRNA | BT125713.1 | - | 6 - 7 | 1934 | 56 |