Assay Design
Gene Transcripts
- Gene Symbol
- egg
- Entrez Gene ID
-
37962
- Gene Name
- eggless
- Gene Aliases
-
AAF47268 Dm, AAM70794, BcDNA:AT13877, CG12196, CG30422, CG30426, Dm-setdb1, DmSETDB1, DmSetdb1, Dmel\CG12196, Dmel_CG12196, Dmsetdb1, EGG, Egg, Eggless, SETDB1, SetDB1, Setdb1, dEset, dSETDB1, dSetDB1, dSetdb1, dsetdb1
- Chromosome Location
-
Chr.2R: 24775534 - 24779901 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086908
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138122.5 |
NP_611966.3 |
3 - 4 |
1433 |
73 |
GenBank mRNA |
BT023947.1 |
- |
3 - 4 |
1421 |
73 |
View Full Product Details