Assay Design



Gene Transcripts

Gene Symbol
emp
Entrez Gene ID
37999
Gene Name
epithelial membrane protein
Gene Aliases
CG2727, Dmel\CG2727, Dmel_CG2727, Emp
Chromosome Location
Chr.2R: 24976456 - 24986070 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010435
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299943.1 NP_001286872.1 4 - 5 715 81
NM_001299944.1 NP_001286873.1 4 - 5 1144 81
NM_079135.4 NP_523859.2 4 - 5 734 81
NM_166702.3 NP_726504.2 4 - 5 701 81
NM_166703.3 NP_726505.1 5 - 6 914 81

View Full Product Details