Assay Design



Gene Transcripts

Gene Symbol
Actn
Entrez Gene ID
31166
Gene Name
alpha actinin
Gene Aliases
ACTN, CG4376, CT14163, CT14232, Dmel\CG4376, Dmel_CG4376, EG:133E12.1, GA17, HM-29, actn, alpha-actinin, alphaActn, fliA, l(1)2Cb, l(1)EA82, y
Chromosome Location
Chr.X: 2023822 - 2042311 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000667
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201603.2 NP_001188532.1 6 - 7 1062 92
NM_001201604.2 NP_001188533.1 6 - 7 1272 92
NM_001272235.1 NP_001259164.1 6 - 7 1267 92
NM_001272236.1 NP_001259165.1 6 - 7 1125 92
NM_001272237.1 NP_001259166.1 6 - 7 1107 92
NM_001272238.1 NP_001259167.1 6 - 7 1122 92
NM_058136.4 NP_477484.2 6 - 7 1267 92
NM_058137.6 NP_477485.1 6 - 7 1330 92
NM_166920.2 NP_726784.1 6 - 7 1267 92
GenBank mRNA AY069615.1 - 6 - 7 1180 92
BT126008.1 - 6 - 7 1268 92

View Full Product Details