Assay Design



Gene Transcripts

Gene Symbol
DIP-alpha
Entrez Gene ID
31322
Gene Name
Dpr-interacting protein alpha
Gene Aliases
32791, CG13019, CG13020, CG32791, Dmel\CG32791, Dmel_CG32791
Chromosome Location
Chr.X: 3507245 - 3571649 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052791
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272288.2 NP_001259217.1 5 - 6 1618 64
NM_001272289.1 NP_001259218.1 3 - 4 654 64
NM_166976.3 NP_726871.1 3 - 4 654 64
GenBank mRNA BT003528.1 - 3 - 4 651 64

View Full Product Details