Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_079961.4 | NP_524700.1 | 9 - 10 | 1683 | 60 |
NM_166982.2 | NP_726877.2 | 8 - 9 | 1634 | 60 | |
GenBank mRNA | AF163775.1 | - | 7 - 8 | 1148 | 60 |
AF220216.1 | - | 9 - 10 | 1421 | 60 | |
BT150204.1 | - | 9 - 10 | 1686 | 60 |