Assay Design



Gene Transcripts

Gene Symbol
bw
Entrez Gene ID
37724
Gene Name
brown
Gene Aliases
CG17632, Dmel\CG17632, Dmel_CG17632, Pm, Su(w[co2]), Su(w[coJ]), ptm
Chromosome Location
Chr.2R: 23527805 - 23538499 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000241
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299840.1 NP_001286769.1 3 - 4 1208 70
NM_079100.3 NP_523824.1 3 - 4 1208 70

View Full Product Details