Assay Design



Gene Transcripts

Gene Symbol
SerT
Entrez Gene ID
37895
Gene Name
Serotonin transporter
Gene Aliases
CG4545, DmSERT, DmSerT, Dmel\CG4545, Dmel_CG4545, SERT, Sert, dSERT, dSERT1, dSerT, dSert, dmSERT, dsert, serT
Chromosome Location
Chr.2R: 24404595 - 24411074 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010414
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299910.1 NP_001286839.1 6 - 7 2421 62
NM_001382130.1 NP_001369117.1 6 - 7 1879 62
NM_079122.3 NP_523846.2 6 - 7 1879 62
GenBank mRNA AY071023.1 - 6 - 7 1876 62
U04809.1 - 5 - 6 1422 62

View Full Product Details