Assay Design
Gene Transcripts
- Gene Symbol
- LysX
- Entrez Gene ID
-
38122
- Gene Name
- Lysozyme X
- Gene Aliases
-
BcDNA:LP09906, CG9120, Dmel\CG9120, Dmel_CG9120, lysX
- Chromosome Location
-
Chr.3L: 1194739 - 1195302 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004431
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079157.2 |
NP_523881.1 |
1 - 1 |
422 |
87 |
GenBank mRNA |
AY119081.1 |
- |
1 - 1 |
422 |
87 |
View Full Product Details