Assay Design



Gene Transcripts

Gene Symbol
Or63a
Entrez Gene ID
38354
Gene Name
Odorant receptor 63a
Gene Aliases
63B.1, 63a, CG9969, DOR28, Dmel\CG9969, Dmel_CG9969, Or63aA
Chromosome Location
Chr.3L: 2961534 - 2963788 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035382
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169860.1 NP_001163331.1 5 - 6 1122 83
NM_079171.4 NP_523895.2 6 - 7 1049 83

View Full Product Details