Assay Design



Gene Transcripts

Gene Symbol
mas
Entrez Gene ID
38499
Gene Name
masquerade
Gene Aliases
CG15002, Dmel\CG15002, Dmel_CG15002, Mas, SPH79, c-SPH79, cSPH79
Chromosome Location
Chr.3L: 4161666 - 4167464 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011653
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274479.1 NP_001261408.1 5 - 6 904 59
NM_079195.3 NP_523919.1 5 - 6 907 59
GenBank mRNA BT001597.1 - 5 - 6 904 59

View Full Product Details