Assay Design
Gene Transcripts
- Gene Symbol
- ndl
- Entrez Gene ID
-
38738
- Gene Name
- nudel
- Gene Aliases
-
CG10129, Dmel\CG10129, Dmel_CG10129, Ndl, SP89, mat(3)2, mel(3)5, nud
- Chromosome Location
-
Chr.3L: 6593938 - 6603366 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002926
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079223.2 |
NP_523947.2 |
3 - 4 |
2246 |
78 |
View Full Product Details