Assay Design



Gene Transcripts

Gene Symbol
Neos
Entrez Gene ID
38795
Gene Name
Neosin
Gene Aliases
BcDNA.HL05936, BcDNA:HL05936, CG8614, Dmel\CG8614, Dmel_CG8614, l(3)L2249
Chromosome Location
Chr.3L: 7241628 - 7243139 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024542
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274573.1 NP_001261502.1 1 - 2 479 77
NM_079231.4 NP_523955.1 1 - 2 668 77
GenBank mRNA BT004827.1 - 1 - 2 593 77

View Full Product Details