Assay Design
Gene Transcripts
- Gene Symbol
- mei-P22
- Entrez Gene ID
-
38797
- Gene Name
- meiotic P22
- Gene Aliases
-
CG14827, Dmel\CG14827, Dmel_CG14827, MEI-P22, Mei-P22
- Chromosome Location
-
Chr.3L: 7244388 - 7245383 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0016036
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079232.1 |
NP_523956.1 |
1 - 1 |
948 |
88 |
GenBank mRNA |
BT100073.4 |
- |
1 - 1 |
948 |
88 |
View Full Product Details