Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169899.2 | NP_001163370.1 | 1 - 2 | 370 | 83 |
NM_001274581.1 | NP_001261510.1 | 1 - 2 | 370 | 83 | |
NM_079234.4 | NP_523958.2 | 1 - 2 | 370 | 83 | |
GenBank mRNA | AY071599.1 | - | 1 - 2 | 336 | 83 |