Assay Design



Gene Transcripts

Gene Symbol
qm
Entrez Gene ID
38816
Gene Name
quemao
Gene Aliases
CG8593, Dmel\CG8593, Dmel_CG8593, GGPPS, GGPPS/qm, Qm, bro2
Chromosome Location
Chr.3L: 7348313 - 7353622 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0019662
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169899.2 NP_001163370.1 2 - 3 514 76
NM_079234.4 NP_523958.2 2 - 3 514 76
GenBank mRNA AY071599.1 - 2 - 3 480 76

View Full Product Details