Assay Design



Gene Transcripts

Gene Symbol
Hn
Entrez Gene ID
38871
Gene Name
Henna
Gene Aliases
CG7399, DTPH, DTPHu, Dmel\CG7399, Dmel_CG7399, PAH, Pah, TPH, TRH, TpH, Tph, Trh, bu, pah
Chromosome Location
Chr.3L: 7760453 - 7763166 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001208
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274613.1 NP_001261542.1 1 - 2 110 59
NM_079239.4 NP_523963.2 1 - 2 208 59
GenBank mRNA AY069306.1 - 1 - 2 94 59

View Full Product Details