Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014573.2 | NP_001014573.1 | 2 - 3 | 1129 | 64 |
NM_001274613.1 | NP_001261542.1 | 3 - 4 | 1039 | 64 | |
NM_001274614.1 | NP_001261543.1 | 4 - 5 | 1333 | 64 | |
NM_079239.4 | NP_523963.2 | 3 - 4 | 1137 | 64 | |
GenBank mRNA | AY069306.1 | - | 3 - 4 | 1023 | 64 |
BT125933.1 | - | 4 - 5 | 1317 | 64 |