Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274686.1 | NP_001261615.1 | 9 - 10 | 3314 | 67 |
NM_001274687.1 | NP_001261616.1 | 9 - 10 | 3314 | 67 | |
NM_079267.4 | NP_523991.2 | 8 - 9 | 1974 | 67 | |
NM_168321.3 | NP_729461.1 | 9 - 10 | 3314 | 67 |