Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274695.1 | NP_001261624.1 | 2 - 3 | 1542 | 84 |
NM_001300070.1 | NP_001286999.1 | 2 - 3 | 1637 | 84 | |
NM_079268.3 | NP_523992.2 | 2 - 3 | 1542 | 84 | |
GenBank mRNA | AY071433.1 | - | 2 - 3 | 1533 | 84 |