Assay Design



Gene Transcripts

Gene Symbol
Ufd1-like
Entrez Gene ID
39254
Gene Name
Ubiquitin fusion-degradation 1-like
Gene Aliases
CG6233, Dmel\CG6233, Dmel_CG6233, UFD1, Udf1, UfD1-like, Ufd-1, Ufd1, Ufd1 like, Ufd1L, anon-WO0118547.405, dUFD1L, dUfd1, ufd1, ufd1-like
Chromosome Location
Chr.3L: 11121966 - 11123216 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036136
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079299.4 NP_524023.1 1 - 2 291 93
GenBank mRNA AF228282.1 - 1 - 2 203 93
AY060735.1 - 1 - 2 277 93

View Full Product Details