Assay Design
Gene Transcripts
- Gene Symbol
- byn
- Entrez Gene ID
-
39349
- Gene Name
- brachyenteron
- Gene Aliases
-
Bra, Byn, CG7260, D-TRA, D-Trg, DTrg, Dm-BYN, Dmel\CG7260, Dmel_CG7260, Tra, Trg, apro, byn/apro, dm-Trg, trg
- Chromosome Location
-
Chr.3L: 11927693 - 11935919 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011723
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079307.2 |
NP_524031.2 |
6 - 7 |
2128 |
76 |
View Full Product Details