Assay Design



Gene Transcripts

Gene Symbol
mirr
Entrez Gene ID
39441
Gene Name
mirror
Gene Aliases
CG10601, DH1, De1, De3, Dmel\CG10601, Dmel_CG10601, Group F, IRO-C, Iro, Iro-C, IroC, MIRR, Mirr, Mrr, Sai, cre, crep, group F, iro, iro-C, l(3)69Ca, l(3)69Da, l(3)6D1, l(3)A5-3-42, mir, mrr
Chromosome Location
Chr.3L: 12693639 - 12710249 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014343
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274849.2 NP_001261778.1 1 - 2 713 66
NM_079323.4 NP_524047.2 1 - 2 713 66
NM_168505.3 NP_729818.1 1 - 2 713 66

View Full Product Details