Assay Design
Gene Transcripts
- Gene Symbol
- Syx8
- Entrez Gene ID
-
39847
- Gene Name
- Syntaxin 8
- Gene Aliases
-
CG4109, Dm Syx8, DmSyx8, Dmel\CG4109, Dmel_CG4109, SYX8, Syx
- Chromosome Location
-
Chr.3L: 16586108 - 16587057 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036643
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079389.4 |
NP_524113.1 |
1 - 2 |
624 |
70 |
View Full Product Details