Assay Design
Gene Transcripts
- Gene Symbol
- cyc
- Entrez Gene ID
-
40162
- Gene Name
- cycle
- Gene Aliases
-
BMAL1, Bmal1, CG8727, CYC, CYCLE, Cyc, Cycle, Dmel\CG8727, Dmel_CG8727, MOP3, Mop3, TGO_CYC, bHLHe7, bMAL1, dBMAL, dBMAL1, dBma1, dBmal, dCYC, dbmal, dbmal1
- Chromosome Location
-
Chr.3L: 19813770 - 19815938 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0023094
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079444.3 |
NP_524168.2 |
4 - 5 |
1143 |
79 |
GenBank mRNA |
BT025197.1 |
- |
4 - 5 |
1143 |
79 |
View Full Product Details