Assay Design



Gene Transcripts

Gene Symbol
fln
Entrez Gene ID
40185
Gene Name
flightin
Gene Aliases
CG7445, Dmel\CG7445, Dmel_CG7445, Fln, Ftn, mp27
Chromosome Location
Chr.3L: 19989551 - 19991220 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005633
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275155.1 NP_001262084.1 2 - 3 533 133
NM_079450.4 NP_524174.1 2 - 3 333 133
GenBank mRNA AY060802.1 - 2 - 3 280 133
Z18858.1 - - 292 133

View Full Product Details