Assay Design
Gene Transcripts
- Gene Symbol
- Las
- Entrez Gene ID
-
40259
- Gene Name
- Lipoic acid synthase
- Gene Aliases
-
CG5231, Dmel\CG5231, Dmel_CG5231
- Chromosome Location
-
Chr.3L: 20430605 - 20432361 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0029158
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079459.3 |
NP_524183.2 |
3 - 4 |
639 |
123 |
View Full Product Details