Assay Design
Gene Transcripts
- Gene Symbol
- CSN3
- Entrez Gene ID
-
40308
- Gene Name
- COP9 signalosome subunit 3
- Gene Aliases
-
CG18332, CH3, Csn3, DCH3, Dch3, Dmel\CG18332, Dmel_CG18332, FBgn0027055, dCSN3
- Chromosome Location
-
Chr.3L: 20839096 - 20841576 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027055
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079466.3 |
NP_524190.2 |
8 - 9 |
1434 |
58 |
View Full Product Details