Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001259928.2 | NP_001246857.1 | 8 - 9 | 1517 | 85 |
NM_001275208.1 | NP_001262137.1 | 14 - 15 | 3316 | 85 | |
NM_001275209.1 | NP_001262138.1 | 15 - 16 | 3527 | 85 | |
NM_001275210.1 | NP_001262139.1 | 15 - 16 | 3524 | 85 | |
NM_079470.5 | NP_524194.3 | 15 - 16 | 3527 | 85 |