Assay Design



Gene Transcripts

Gene Symbol
alpha-Cat
Entrez Gene ID
40517
Gene Name
alpha Catenin
Gene Aliases
CG17947, CT39986, Dalpha-Cat, Dalpha-cat, DalphaC, Dalphacat, Dmcatp, Dmel\CG17947, Dmel_CG17947, LD30423p, a-Cat, a-cat, alpha-CAT, alpha-cat, alpha-catenin, alpha-ctn, alphaCAT, alphaCat, alphacat
Chromosome Location
Chr.3L: 23318646 - 23338100 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010215
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300225.1 NP_001287154.1 1 - 2 229 116
NM_079495.3 NP_524219.1 1 - 2 229 116
GenBank mRNA D13964.2 - 1 - 2 192 116

View Full Product Details