Assay Design



Gene Transcripts

Gene Symbol
stj
Entrez Gene ID
36526
Gene Name
straightjacket
Gene Aliases
CG12295, Dmel\CG12295, Dmel_CG12295, alpha[[2]]delta, alpha[[2]]delta-3, dalpha[[2]]delta-3, l(2)k10814
Chromosome Location
Chr.2R: 13805794 - 13819571 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261041
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259375.2 NP_001246304.1 15 - 16 3202 76
NM_137058.4 NP_610902.1 14 - 15 3145 76
GenBank mRNA AY069830.1 - 14 - 15 3465 76

View Full Product Details