Assay Design



Gene Transcripts

Gene Symbol
conv
Entrez Gene ID
36588
Gene Name
convoluted
Gene Aliases
ALS, Als, CG8561, CT24963, Dmel\CG8561, Dmel_CG8561, dALS, l(2)k06507b, tartan/capricious-like
Chromosome Location
Chr.2R: 14270729 - 14275495 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261269
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299487.1 NP_001286416.1 7 - 8 3087 64
NM_137104.3 NP_610948.2 7 - 8 3328 64
GenBank mRNA EU814872.1 - 7 - 8 2887 64

View Full Product Details