Assay Design
Gene Transcripts
- Gene Symbol
- Dh44-R1
- Entrez Gene ID
-
36601
- Gene Name
- Diuretic hormone 44 receptor 1
- Gene Aliases
-
CG8422, DH-R, DH44-R1, DH44R1, DH[[44]]-R1, Dh44R1, DmeCG8422, Dmel\CG8422, Dmel_CG8422, anon-WO0170980.100, anon-WO0170980.101
- Chromosome Location
-
Chr.2R: 14378097 - 14385359 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033932
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137116.3 |
NP_610960.1 |
11 - 12 |
2092 |
64 |
View Full Product Details