Assay Design



Gene Transcripts

Gene Symbol
Lap1
Entrez Gene ID
36670
Gene Name
CG10255-PA
Gene Aliases
CG10255, CT28815, Dmel\CG10255, Dmel_CG10255, dLAP1, dlap1
Chromosome Location
Chr.2R: 14935660 - 14940722 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033984
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202009.2 NP_001188938.1 5 - 6 2787 113
NM_137163.5 NP_611007.1 5 - 6 2770 113
GenBank mRNA BT011365.1 - 5 - 6 2771 113

View Full Product Details