Assay Design
Gene Transcripts
- Gene Symbol
- ey
- Entrez Gene ID
-
43812
- Gene Name
- eyeless
- Gene Aliases
-
CG1464, DPax-6, Dmel\CG1464, Dmel_CG1464, EY, EYEL, Ey, Ey/Pax6, OK107, Pax-6, Pax6, Pax6/ey, eye, l(4)102CDh, l(4)102CDr, l(4)33, pax6
- Chromosome Location
-
Chr.4: 697689 - 721173 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0005558
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079889.3 |
NP_524628.2 |
1 - 2 |
629 |
66 |
View Full Product Details